missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RABIF Recombinant Protein
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH18488 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 5877 |
Molecular Weight (g/mol) | 39.27 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130002
|
Abnova™
H00005877-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140002
|
Abnova™
H00005877-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH18488 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
39.27 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RABIF | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
5877 | |
RABIF (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPAEQPSELVSAEGRNRKAVLCQRCGSRVLQPGTALFSRRQLFLPSMRKKPALSDGSNPDGDLLQEHWLVEDMFIFENVGFTKDVGNIKFLVCADCEIGPIGWHCLDDKNSFYVALERVSHE | |
MSS4/RASGFR3/RASGRF3 | |
RABIF | |
Wheat Germ (in vitro) | |
GST |