Learn More
Thermo Scientific™ R15K, HIV-1 Inhibitory Peptide
For cellular and molecular biology applications
566.00 NOK - 723.00 NOK
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1655.97
Product Type R15K, HIV-1 Inhibitory Peptide
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15447639
SDS View Documents Promotion Details
Thermo Scientific™
J66327.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15457639
SDS View Documents Promotion Details
Thermo Scientific™
J66327.M^
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
HPLC
Solid
R15K, HIV-1 Inhibitory Peptide
Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys
-30°C to -10°C
1655.97
RUO - research use only
Documents
Product Certifications
For Research Use Only