missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone, 98%, Thermo Scientific™

Catalog Number 15457957
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 157636-81-2
Molecular Formula C12H12N2O2
MDL Number MFCD01863583
Quantity 1 g
Sensitivity Air Sensitive
Synonym r-4-1h-indol-3-yl methyl oxazolidin-2-one, r---4-1h-indol-3-ylmethyl-2-oxazolidinone, 4r-4-1h-indol-3-ylmethyl-1,3-oxazolidin-2-one, pubchem12405, 4r-4-1h-indol-3-ylmethyl-2-oxazolidinone, r---4-3-indolylmethyl-2-oxazolidinone, 2-oxazolidinone,4-1h-indol-3-ylmethyl-, 4r, 4r-4-1h-indol-3-yl methyl-1,3-oxazolidin-2-one
InChI Key LGXHODFXCOIGGJ-SECBINFHSA-N
SMILES C1C(NC(=O)O1)CC2=CNC3=CC=CC=C32
IUPAC Name (4R)-4-(1H-indol-3-ylmethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 216.24
PubChem CID 16213659
Formula Weight 216.24
Percent Purity 98%
Chemical Name or Material (R)-(-)-4-(3-Indolylmethyl)-2-oxazolidinone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'