missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (R)-(-)-2-Methyl-2,4-pentanediol, 98+%

Brand:  Alfa Aesar™ L14523.03

Additional Details : CAS Number : 99210-90-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11393566

  • 942.00 NOK / 1g


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Applications
(R)-(-)-2-Methyl-2,4-pentanediol is used as a chiral reagent. Also used as intermediate for pharmaceutical.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

99210-90-9
118.176
SVTBMSDMJJWYQN-RXMQYKEDSA-N
5288845
CC(CC(C)(C)O)O
C6H14O2
MFCD00064275
4r-2-methylpentane-2,4-diol, r---2-methyl-2,4-pentanediol, mrd, 2-methylpentan-2,4-diol, r-2-methyl-2,4-pentanediol, 1hty, r-mpd, 4-methyl-2,4-pentanediole, r---2-methyl-2,4-pentandiol, .+/-.-2-methyl-2,4-pentanediol
(4R)-2-methylpentane-2,4-diol

Specifications

(R)-(-)-2-Methyl-2,4-pentanediol
93°C (199°F)
1.4275
197°C
99210-90-9
MFCD00064275
4r-2-methylpentane-2,4-diol, r---2-methyl-2,4-pentanediol, mrd, 2-methylpentan-2,4-diol, r-2-methyl-2,4-pentanediol, 1hty, r-mpd, 4-methyl-2,4-pentanediole, r---2-methyl-2,4-pentandiol, .+/-.-2-methyl-2,4-pentanediol
SVTBMSDMJJWYQN-RXMQYKEDSA-N
(4R)-2-methylpentane-2,4-diol
5288845
≥98%
0.92
−21° (Neat)
Hygroscopic
1g
C6H14O2
6051650
Fully miscible with water.
CC(CC(C)(C)O)O
118.176
118.18
Videos
SDS Documents