missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, 97%, 99+% ee, Thermo Scientific™

Catalog Number p-3777716
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
64265-77-6
C8H11BrClN
236.54
MFCD02259386
BQCAANUXMMQVAY-UHFFFAOYNA-N
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1
2734642
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride
Cl.CC(N)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
64265-77-6
C8H11BrClN
236.54
MFCD02259386
BQCAANUXMMQVAY-UHFFFAOYNA-N
r-1-4-bromophenyl ethanamine hydrochloride, r-+-1-4-bromophenyl ethylamine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride, 1r-1-4-bromophenyl ethylamine, chloride, r-1-4-bromophenyl ethanamine hcl, r-1-4-bromophenyl ethylamine hcl, r-1-4-bromophenyl ethanaminehydrochloride, r-1-4-bromophenyl ethylaminehydrochloride, 1r-1-4-bromophenyl ethan-1-amine hydrochloride, 1r-1-4-bromophenyl ethanamine hydrochloride 1:1
2734642
(1R)-1-(4-bromophenyl)ethanamine;hydrochloride
Cl.CC(N)C1=CC=C(Br)C=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 238°C to 245°C
Color White to Yellow
Enantiomeric Excess 99.0% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with alcohols
Formula Weight 236.54
Percent Purity 97%,99+% ee
Physical Form Powder
Chemical Name or Material (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, ≥99%
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'