missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Quinoline-3-carboxylic acid, 98%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | ∽283°C (decomposition) |
Quantity | 1 g |
Beilstein | 126542 |
Formula Weight | 173.17 |
Percent Purity | 98% |
Chemical Name or Material | Quinoline-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'