missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 66°C to 70°C |
Quantity | 5 g |
Beilstein | 113061 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 157.17 |
Percent Purity | 97% |
Chemical Name or Material | Quinoline-2-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'