Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Ideally suited for use with volumetric flasks.
Brand:  Quickfit TRF697
1054.50 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.33000kg
 View more versions of this product
Product Code. 12471788
  • 1110.00 NOK / Pack of 10
Estimated Shipment: 17-07-2024
Description
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
Specifications
Specifications
Hollow-Blown Glass
10/Pk.
Volumetric flasks
29/32
Glass
NS 29/32