Learn More
Pyruvic aldehyde, 35-45 wt% solution in water, Thermo Scientific Chemicals
Catalog Number p-189912
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
78-98-8, 7732-18-5
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
78-98-8, 7732-18-5
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -20.0°C
Color Brown-Yellow to Yellow
Density 1.2000g/mL
pH 1 to 3
Boiling Point >72.0°C
Flash Point >71°C
Packaging Glass bottle
Linear Formula CH3COCHO
Refractive Index 1.4209
Quantity 100 g
Beilstein 01, 762
Merck Index 15, 6153
Specific Gravity 1.2
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, benzene and dmso
Formula Weight 72.06
Percent Purity 35-45 wt.%
Physical Form Solution
Chemical Name or Material Pyruvic aldehyde, 35-45 wt.%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyruvic aldehyde, 35-45 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.