Learn More
Pyridinium bromide perbromide, 90+%, technical, Thermo Scientific Chemicals
Catalog Number p-189701
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
39416-48-3
C5H6Br3N
319.83
MFCD00013223
Pyridine hydrobromide, compound with bromine
39416-48-3
C5H6Br3N
319.83
MFCD00013223
Pyridine hydrobromide, compound with bromine
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Melting Point 127°C to 133°C
Color Brown to Orange-Red
Infrared Spectrum Authentic
Assay Percent Range 90% min. (Iodometry)
Packaging Glass bottle
Quantity 100 g
Beilstein 20, V,5, 181
Fieser 01,967; 05,568; 06,499; 09,399; 10,333; 14,268; 15,276
Merck Index 15, 8085
Solubility Information Solubility in water: decomposes. Other solubilities: sparingly soluble in acetic acid
Formula Weight 319.83
Percent Purity 90+%
Grade Technical
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Pyridinium bromide perbromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Pyridinium bromide perbromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.