Learn More
Pulegone, 92%, pract., Thermo Scientific Chemicals
Catalog Number p-3779529
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
89-82-7
C10H16O
152.24
MFCD00063000
NZGWDASTMWDZIW-QMMMGPOBSA-N
--pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
638012
CHEBI:81226
(5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
CC1CCC(=C(C)C)C(=O)C1
89-82-7
C10H16O
152.24
MFCD00063000
NZGWDASTMWDZIW-QMMMGPOBSA-N
--pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one
638012
CHEBI:81226
(5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one
CC1CCC(=C(C)C)C(=O)C1
Pulegone, CAS # 89-82-7, is a compound found in the oils of certain plant species including peppermint, catnip, and pennyroyal.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations
Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9310g/mL
Boiling Point 97°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.488
Quantity 100 g
Beilstein 07, 81
Merck Index 15, 8046
Specific Gravity 0.931
Specific Rotation Condition +21° (20°C neat)
Specific Rotation + 21
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,chloroform,diethyl ether
Formula Weight 152.24
Percent Purity 92%
Grade Practical
Physical Form Liquid
Chemical Name or Material Pulegone