Learn More
PTPN3 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-18412JF525
Product Code. 30171663
  • 5565.00 NOK / 0.10mL
Description
PTPN3 Polyclonal antibody specifically detects PTPN3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTPN3
Polyclonal
50mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
Janelia Fluor 525
DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3
Recognizes region between residues Between between 175 and 225..
0.1 mL
Immunology, Signal Transduction
5774
Store at 4°C in the dark.