Learn More
Merck Biopore Membrane Filter Roll, Hydrophilic PTFE
Brand:  Merck BGCM00010
Additional Details : Weight : 0.01000kg
Product Code. 10448213
  • 16155.00 NOK / Each
Estimated Shipment: 16-05-2024
Description
Biopore Membrane Filter roll, Hydrophilic PTFE
Specifications
Specifications
Non-sterile
50 mL
Biopore Membrane Filter Roll
1 Ea.
PTFE
0.4 μm
12 mm