Learn More
Pseudothiohydantoin, 97%, Thermo Scientific Chemicals
Catalog Number p-7025789
Click to view available options
Quantity:
5 g
25 g
100 g
556-90-1
C3H4N2OS
116.138
MFCD00003186
HYMJHROUVPWYNQ-UHFFFAOYSA-N
pseudothiohydantoin, 2-aminothiazol-4 5h-one, 2-imino-4-thiazolidinone, 4 5h-thiazolone, 2-amino, usaf dm-1, 2-imino-4-thiazolidone, 2-amino-2-thiazolin-4-one, usaf be-4-5, 2-amino-4-oxo-thiazole, 2-imino-thiazolidin-4-one
11175
2-amino-1,3-thiazol-4-one
C1C(=O)N=C(S1)N
556-90-1
C3H4N2OS
116.138
MFCD00003186
HYMJHROUVPWYNQ-UHFFFAOYSA-N
pseudothiohydantoin, 2-aminothiazol-4 5h-one, 2-imino-4-thiazolidinone, 4 5h-thiazolone, 2-amino, usaf dm-1, 2-imino-4-thiazolidone, 2-amino-2-thiazolin-4-one, usaf be-4-5, 2-amino-4-oxo-thiazole, 2-imino-thiazolidin-4-one
11175
2-amino-1,3-thiazol-4-one
C1C(=O)N=C(S1)N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point ∼240°C (decomposition)
Quantity 5 g
Formula Weight 116.14
Percent Purity 97%
Chemical Name or Material Pseudothiohydantoin