Learn More
Trajan Scientific and Medical™ ProteCo™ PEEK Stainless Steel Unions
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Stainless Steel Unions offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 222006
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15269213
  • 1335.00 NOK / Pack of 2
Estimated Shipment
15-07-2020
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
Size: 1/32 in. to 0.36 mm
Stainless Steel
PEEK Stainless Steel Unions 1/32 in. to 0.36mm