missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Propargyl methacrylate, 98%, stab. with 250ppm 4-methoxyphenol

Brand:  Alfa Aesar™ L11556.18

Additional Details : CAS Number : 13861-22-8 Weight : 0.05000kg Transport : UN number : 3272 Chem class : 3 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 11392487

  • 4229.00 NOK / 50g


In stock, ready to ship: 6
Add to basket

Description

Description

Applications
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

13861-22-8
124.139
PZAWASVJOPLHCJ-UHFFFAOYSA-N
83778
CC(=C)C(=O)OCC#C
C7H8O2
MFCD00048123
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
prop-2-ynyl 2-methylprop-2-enoate

Specifications

Propargyl methacrylate
48°C (118°F)
149°C to 151°C
stabilized with 250ppm 4-methoxyphenol
C7H8O2
UN3272
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
PZAWASVJOPLHCJ-UHFFFAOYSA-N
prop-2-ynyl 2-methylprop-2-enoate
83778
98%
0.986
1.45
50g
13861-22-8
MFCD00048123
906850
Immiscible with water.
CC(=C)C(=O)OCC#C
124.139
124.14
Videos
SDS Documents