missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (44D9) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-14817JF635
This item is not returnable.
View return policy
Description
Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
| Procalcitonin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
| 0.2 mL | |
| Cancer, Cellular Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 796 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Sandwich ELISA, Immunocytochemistry/Immunofluorescence | |
| 44D9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction