missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PRDM16 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0986
Additional Details : Weight : 0.01000kg
Description
PRDM16 Synthetic Peptide, PEP-0986, from Invitrogen.
This peptide corresponds to 17 amino acids near the carboxy terminus of human PRDM16.PEP-0986 can be used as a blocking peptide with polyclonal antibody PA5-20872.Specifications
63976 | |
purified | |
Human PRDM16 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9HAZ2 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
PRDM16 | |
0.02% sodium azide | |
50 μg | |
PBS |