missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PPARG (Human) Recombinant Protein

Human PPARG full-length ORF ( AAH06811, 1 a.a. - 477 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005468-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189381

  • 3655.00 NOK / 10µg
Estimated Shipment: 23-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MTMVDTEMPFWPTNFGISSVDLSVMEDHSHSFDIKPFTTVDFSSISTPHYEDIPFTRTDPVVADYKYDLKLQEYQSAIKVEPASPPYYSEKTQLYNKPHEEPSNSLMAIECRVCGDKASGFHYGVHACEGCKGFFRRTIRLKLIYDRCDLNCRIHKKSRNKCQYCRFQKCLAVGMSHNAIRFGRMPQAEKEKLLAEISSDIDQLNPESADLRALAKHLYDSYIKSFPLTKAKARAILTGKTTDKSPFVIYDMNSLMMGEDKIKFKHITPLQEQSKEVAIRIFQGCQFRSVEAVQEITGYAKSTPGFVNLDLNDQVTLLKYGVHEIIYTMLASLMNKDGVLISEGQGFMTREFLKSLRKPFGDFMEPKFEFAVKFNALELDDSDLAIFIAVIILSGDRPGLLNVKPIEDIQDNLLQALELQLKLNHPESSQLFAKLLQKMTDLRQIVTEHVQLLQVIKKTETDMSLHPLLQEIYKDLY
Specifications

Specifications

AAH06811
peroxisome proliferator-activated receptor gamma
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPARG
GST
5468
Wheat germ expression system
10 μg
CIMT1, NR1C3, PPARG1, PPARG2, PPARgamma
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers