Learn More
Abnova™ PPARG/CEN3q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0216.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CIMT1,NR1C3,PPARG1,PPARG2,PPARgamma | |
5468 | |
200 μL |
Tube | |
peroxisome proliferator-activated receptor gamma | |
PPARG | |
DNA/RNA | |
Liquid |