missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetraphenylborate, 97%, Thermo Scientific Chemicals

Catalog Number p-7052681
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3244-41-5
C24H20BK
358.332
MFCD03093877
MAMCUZQNCMYPCY-UHFFFAOYSA-N
potassium tetraphenylborate, potassium tetraphenylboron, potassium tetraphenylboride, potassium tetrakis phenyl borate, potassium tetraphenyl boron, potassium tetraphenylborate 97, potassium tetraphenylborate 1-
23665558
potassium;tetraphenylboranuide
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]
This item is not returnable. View return policy
3244-41-5
C24H20BK
358.332
MFCD03093877
MAMCUZQNCMYPCY-UHFFFAOYSA-N
potassium tetraphenylborate, potassium tetraphenylboron, potassium tetraphenylboride, potassium tetrakis phenyl borate, potassium tetraphenyl boron, potassium tetraphenylborate 97, potassium tetraphenylborate 1-
23665558
potassium;tetraphenylboranuide
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[K+]

It is an active pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula KB(C6H5)4
Solubility Information Low solubility in water. Soluble in organic solvents.
Formula Weight 358.33
Percent Purity 97%
Chemical Name or Material Potassium tetraphenylborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'