missing translation for 'onlineSavingsMsg'
Learn More

Potassium peroxydisulfate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog Number p-7080255
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
7727-21-1
K2O8S2
270.309
MFCD00011386
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
This item is not returnable. View return policy
7727-21-1
K2O8S2
270.309
MFCD00011386
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Potassium persulfate is a powerful oxidant, commonly used to initiate polymerizations. Potassium Persulphate used for bleaching and textile desizing, as an oxidizing agent and antiseptic, in purification of ammonium sulfate, and in the manufacture of soap and pharmaceuticals. It is also used as al laboratory oxidant and photography chemical. It is a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C (dec.)
Color White
Physical Form Crystalline
Quantity 250 g
Linear Formula K2S2O8
UN Number UN1492
Merck Index 14,7656
Solubility Information It is highly soluble in water. It is insoluble in alcohol.
Formula Weight 270.31
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Density 2.477 g/mL
Chemical Name or Material Potassium peroxydisulfate, Low nitrogen
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'