compliance-icons
Potassium Permanganate Solution, 0.02M, Fisher Chemical™
BIOPHARMA
Product Code. 12930764 Shop All Fisher Chemical Products
100 mL, 100mL
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 12930764
Brand: Fisher Chemical SOL 119 100
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 0.02mol/L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number1155396 Certificate TypeCertificate of Analysis Date26/09/2025 Product CodeSOL 119 100
Lot Number1146647 Certificate TypeCertificate of Analysis Date21/08/2025 Product CodeSOL 119 100
Lot Number1122939 Certificate TypeCertificate of Analysis Date13/06/2025 Product CodeSOL 119 100
Lot Number1072196 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeSOL 119 100
Lot Number1066954 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeSOL 119 100
compliance-icons
Product Identifier
  • Potassium permanganate 0.2M Volumetric Solution (SOL 119)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Potassium Permanganate Solution, 0.02M, Fisher Chemical™ >