missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide solution, 1M EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Concentration | 1M |
CAS | 1310-58-3, 7732-18-5 |
Packaging | Bottle |
Quantity | 1000 mL |
Molecular Formula | HKO |
MDL Number | MFCD00003553 |
UN Number | 1814 |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
SMILES | [OH-].[K+] |
Show More |
Safety and Handling
Product Identifier
- Potassium hydroxide 1M. Volumetric Solution European Pharmacopoeia Ref: 3009100
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'