missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.0145M, Conductivity Standard, Thermo Scientific Chemicals

Catalog Number p-7052159
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
[Cl-].[K+]
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
[Cl-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 0.0145M
Quantity 500 mL
IUPAC Name potassium chloride
Formula Weight 74.55
Odor Odorless
Physical Form Liquid
Chemical Name or Material Potassium chloride Conductivity Standard, 2000μmho at 25°C

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'