missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048546
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
[K+].[Br-]

Potassium bromide is used in a variety of applications such as spectroscopy, infrared transmission, as a laboratory reagent, and for the removal of peripheral membrane proteins. It is also used in photosensitive chemicals, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula KBr
Merck Index 14,7618
Solubility Information Soluble in water. Slightly soluble in alcohol,glycerol and ether.
IUPAC Name potassium;bromide
Formula Weight 119.01
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75 g/mL
Assay (metals basis)
Chemical Name or Material Potassium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'