Learn More
Potassium Acetate (Colorless Crystals or Powder), Fisher BioReagents
Brand: Fisher Bioreagents BP364-500
Additional Details : CAS Number : 127-08-2 Weight : 0.55000kg
Description
Poly Bottle; 500g
Specifications
Potassium Acetate | |
127-08-2 | |
7.0 to 8.0 | |
500 g | |
C2H3KO2 | |
15, 7726 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Pass Test |
292°C | |
White | |
Solid | |
≥99.0 % | |
CH3COOK | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
≥99.0% | |
Poly Bottle |