Learn More
Haldenwanger™ Porcelain Spoon Spatula
177.02 NOK - 496.00 NOK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10169150
|
Haldenwanger™
74/2 |
121 mm |
177.02 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10508261
|
Haldenwanger™
74/6 |
210 mm |
322.00 NOK
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10508071
|
Haldenwanger™
74/4 |
170 mm |
225.00 NOK
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10028430
|
Haldenwanger™
74/8 |
305 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
1 Pk. | |
Spoon Spatula | |
Porcelain |