Learn More
Haldenwanger™ Porcelain Incincerating Dish
Brand: Haldenwanger™ 33/2
Additional Details : Weight : 0.27000kg
Description
Order Info
CapacityMetric
Specifications
15 mL | |
Round | |
25 mm |
Porcelain | |
Incinerating Dish | |
40 mm |