Learn More
Kartell™ Polypropylene Micropipet Tips
Designed with high clarity and excellent water-repellent properties to keep sample retention within the tip to a minimum. Kartell™ Polypropylene Micropipet Tips feature precise centering of the orifice at the end of the tip to guarantee accurate delivery of the sample.
Brand:  Kartell™ 0094300
Additional Details : Weight : 0.80000kg
Product Code. 12523377
  • 3550.00 NOK / Pack of 1000
Estimated Shipment: 18-04-2024
Description
  • Cadmium-free polypropylene construction provides high clarity and water-repellent properties
  • Specially designed for Kartell automatic pipets, but deliver excellent results with most available micropipet brands 
  • Autoclavable to 121°C for 20 minutes
  • Precision delivery of sample, minimal sample retention with tip 
  • Packaging selection satisfies a variety of needs:
    • bulk bag
    • packed in racks
    • refills for reloading racks