Learn More
Kartell™ Plastilab™ Polypropylene 4-Way Connectors
For use with plastic or rubber tubing.
Brand:  Kartell™ 7141000
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 12519776
Quantity Price
1 261.00 NOK / Pack of 10
10 247.95 NOK / Pack of 10
Estimated Shipment: 15-10-2021
Add to basket
Description
Description
  • Sturdy shape provides strength
  • Chemical and thermal resistant
Specifications
Specifications
CE IVD
3.5 mm
Polypropylene
1.5mm Bore
Natural
Autoclavable
10/Pk.
4-way cross