Learn More
Brady™ Polyester Labels for the BBP33/i3300 Printer
Click to view available options
Color:
Blue, White
Green, White
Orange, White
Pink, White
Red, White
White
Yellow, White
Length (Metric):
9.53 mm
12.70 mm
15.24 mm
Unit Size:
Pack of 1500
Pack of 2500
Description
- The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
- Thanks to subsurface printing, the label's color code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifica
Specifica
| Certifications/Compliance | Compliant with Directive 2011/65/EU and 2015/863 |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | Yellow, White |
| Finish | Gloss |
| Format | Roll |
| Material | Polyester |
| Printable Sides | Front |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto