Learn More
3M™ Faceshield, Polycarbonate, Clear, 5F-11, 10 ea/Case
1270.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Description
- 3M™ Faceshield certified to EN 166:2001
- Provides protection against secondary welding light
- Anti-fog and anti-scratch coated, optical class 1, clear curved polycarbonate faceshield
- Designed to fit 3M™ Hard Hats Series G3000 or G22 with 3M™ V5 Faceshield Mounting system
- Snap in feature makes it quick and easy to connect to the Faceshield Holder
- Medium energy impact, Grade B
- Optical class 1 visor, low energy impact, B
Specifications
Thickness (Metric) | 1.5 mm |
Product Type | Faceshield |
Material | Polycarbonate |