missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | POLR1D |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899853
|
Proteintech
12254-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16889853
|
Proteintech
12254-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a component of the RNA polymerase I and RNA polymerase III complexes, which function in the synthesis of ribosomal RNA precursors and small RNAs, respectively. Mutations in this gene are a cause of Treacher Collins syndrome (TCS), a craniofacial development disorder. Alternative splicing results in multiple transcript variants.Specifications
POLR1D | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P97304, Q9Y2S0 | |
20018, 360762, 51082 | |
POLR1D Fusion Protein Ag2904 | |
Primary | |
-20°C | |
POLR1D |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AC19, hRPA19, POLR1C, POLR1D, RPA16, RPA9, RPAC2, RPC16, RPO1 3 | |
Polr1d | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |