Learn More
Motic™ Simple Add-on Analyzer Filter
Restricts detected light to refracted light and must be used in conjunction with Motic™ Simple Add-on Polarizer. Motic™ Simple Add-on Analyzer Filter is for use with BA210 or BA310 microscope.
Brand: Motic™ 1101001900581
Additional Details : Weight : 0.80000kg
Description
- For use with BA210/310 microscope
- Requires use with Polarizer
- Produces refracted light while polarizer illuminates the specimen
Order Info
ForUseWithEquipment
Specifications
Analyzer Filter | |
Restricting detected light | |
1 Ea. |
Requires use with Polarizer Filter | |
BA310 Series Microscope |