Learn More
BRAND™ PMP Class A Measuring Flask
Product Code. 10465243 Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 2
Product Code. 10465243
Brand: BRAND™ 36178
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PMP flask with polypropylene stopper
  • Error limits according to class A, DIN EN ISO 1042
  • Includes one batch certificate
  • Calibrated to contain (TC, In)
  • Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
  • TRUSTED_SUSTAINABILITY
    Specifications
    Capacity (Metric) 250 mL
    Material PMP
    Includes PP stopper
    Socket Joint Ground joint: NS 19/26
    Tolerance 0.15 mL
    Autoclavable Autoclavable
    Class A