missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension (Od/Id 60.3/57 mm), Angled, For Exhaust Air Ducting For U750

Use these tube extensions with Memmert ovens. Memmert&trade Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.

Brand:  Memmert™ U750B29719

Additional Details : Weight : 0.01100kg

Product Code. 16376447

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
  • Angled
  • Only models U, I, S
Specifications

Specifications

Exhaust Air Ducting
Plug-in Tube Extension
2.2 x 2.4 in. (I.D. x O.D.)
57 x 60.3 mm (I.D. x O.D.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers