Learn More
Alfa Aesar™ Platinum wire, 0.127mm (0.005 in.) dia., For ISA Type R or S Standard Grade Thermocouple
For ISA Type R or S Standard Grade Thermocouple
Brand: Alfa Aesar™ 010289.G1
Additional Details : CAS Number : 7440-06-4 Weight : 0.01000kg
Quantity | 1m |
---|
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
Platinum wire | |
1m | |
7440-06-4 | |
MFCD00011179 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
195.08 | |
CHEBI:33400 |
≈0.27g/m | |
0.127mm (0.005 in.) dia. | |
Pt | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
Odorless | |
23939 | |
Wire, ≈0.27g/m |