missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7076402
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy
Pt:Rh; 80:20 wt%
MFCD00198186
This item is not returnable. View return policy
Pt:Rh; 80:20 wt%
MFCD00198186

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Tube
Quantity 25 x 25 mm
Percent Purity 99.9%
Chemical Name or Material Platinum Rhodium foil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'