Learn More
Macherey-Nagel™ Plastic Syringe
Graduated plastic syringe, 1mL capacity, for use with thin layer chromatography (TLC) applications.
Brand: Macherey-Nagel™ 814104
Additional Details : Weight : 0.01000kg
Specifications
Plastic (Barrel) | |
Syringe | |
Capillaries |
Thin layer chromatography (TLC) | |
1 mL | |
Yes |