Learn More
Microspec™ Plastic Culture Loop
Click to view available options
Volume:
1 μL
5 μL
10 μL
Unit Size:
Pack of 12000
Description
- 12,000 Plastic Culture Loops
- Microseal Pouch of 20
- 200 mm length
- Sterile EtO
Specifications
Specifications
| Disposable | Single-use |
| Volume | 10 μL |
| Length (English) | 7.8 in. |
| Length (Metric) | 200 mm |
| Material | Plastic |
| Sterility | Sterile |
| Size | Standard |
| Color | Blue |
| Product Type | Inoculating Loop |
| Autoclavable | Not Autoclavable |