missing translation for 'onlineSavingsMsg'
Learn More

Pivaloyl hydrazide, 98+%, Thermo Scientific Chemicals

Catalog Number p-3213062
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
42826-42-6
C5H12N2O
116.16
MFCD01310839
OARJXUPBZNUYBG-UHFFFAOYSA-N
2,2-dimethylpropionic acid hydrazide, pivalic acid hydrazide, pivalohydrazide, 2,2-dimethylpropanohydrazide, pivaloylhydrazine, 2,2-dimethyl-propionic acid hydrazide, propanoic acid, 2,2-dimethyl-, hydrazide, pivalic acid, hydrazide, 2,2-dimethylpropionicacidhydrazide, trimethylacethydrazide
545147
2,2-dimethylpropanehydrazide
CC(C)(C)C(=O)NN
This item is not returnable. View return policy
42826-42-6
C5H12N2O
116.16
MFCD01310839
OARJXUPBZNUYBG-UHFFFAOYSA-N
2,2-dimethylpropionic acid hydrazide, pivalic acid hydrazide, pivalohydrazide, 2,2-dimethylpropanohydrazide, pivaloylhydrazine, 2,2-dimethyl-propionic acid hydrazide, propanoic acid, 2,2-dimethyl-, hydrazide, pivalic acid, hydrazide, 2,2-dimethylpropionicacidhydrazide, trimethylacethydrazide
545147
2,2-dimethylpropanehydrazide
CC(C)(C)C(=O)NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65°C to 67°C
Color White
Boiling Point 95°C to 110°C (1.0 mbar)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC(O)NHNH2
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols
Formula Weight 116.16
Percent Purity 98+%
Physical Form Crystals
Chemical Name or Material Pivaloyl hydrazide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pivaloyl hydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'