Learn More
Pirmenol hydrochloride, MedChemExpress
Product Code. 16464456
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16464456
Brand: Medchem Express HY100795A
MedChemExpress Pirmenol hydrochloride inhibits IK.ACh by blocking muscarinic receptors. The IC50 of Pirmenol for inhibition of Carbachol-induced IK.ACh is 0.1 μM.
Pirmenol hydrochloride inhibits IK.ACh by blocking muscarinic receptors. The IC50 of Pirmenol for inhibition of Carbachol-induced IK.ACh is 0.1 μM.
Specifications
Chemical Name or Material Pirmenol hydrochloride
Physical Form Solid
CAS 61477-94-9
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H31ClN2O
For Use With (Application) COVID-19-immunoregulation
Synonym Cl-845 (±)-Pirmenol hydrochlorid
Solubility Information DMSO : 125 mg/mL (333.38 mM; Need ultrasonic)
SMILES OC(C1=CC=CC=C1)(CCCN2[C@@H](C)CCC[C@H]2C)C3=CC=CC=N3.Cl[H]
Molecular Weight (g/mol) 374.95
Formula Weight 374.95
Percent Purity 95.63%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.