Learn More
KARTELL Pippette Filter Tip Box with Lid
Description
Empty PP boxes with snap-on lid for pipette filter tips.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Gray |
| Holds | Pipette Tips |
| Material | Polypropylene |
| Sterility | Non-sterile |
| For Use With (Equipment) | Pipette Filter Tips |
| Length (Metric) | 12.52 cm |
| Includes | Box with Lid |
| Product Type | Pipet Box Holder |