Learn More
Piperazine-2-one, 95%, Thermo Scientific Chemicals
Catalog Number p-3779480
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
5625-67-2
C4H9N2O
101.13
MFCD01318687
IWELDVXSEVIIGI-UHFFFAOYSA-O
2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine
231360
piperazin-2-one
O=C1C[NH2+]CCN1
5625-67-2
C4H9N2O
101.13
MFCD01318687
IWELDVXSEVIIGI-UHFFFAOYSA-O
2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine
231360
piperazin-2-one
O=C1C[NH2+]CCN1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 136°C to 140°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 100.12
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Piperazine-2-one
compliance-icons
Product Identifier
  • Piperazine-2-one
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.