missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PIK3CA (Human) Recombinant Protein
Human PIK3CA partial ORF ( NP_006209, 959 a.a. - 1068 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005290-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: DFLIVISKGAQECTKTREFERFQEMCYKAYLAIRQHANLFINLFSMMLGSGMPELQSFDDIAYIRKTLALDKTEQEALEYFMKQMNDAHHGGWTTKMDWIFHTIKQHALN
Specifications
NP_006209 | |
phosphoinositide-3-kinase, catalytic, alpha polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIK3CA | |
GST |
5290 | |
Wheat germ expression system | |
10 μg | |
MGC142161, MGC142163, PI3K, p110-alpha | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |