missing translation for 'onlineSavingsMsg'
Learn More

PIGK Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15151-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884204

  • 3468.15 NOK / 150µL
Estimated Shipment: 15-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the cysteine protease family C13 that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is a member of the multisubunit enzyme, GPI transamidase and is thought to be its enzymatic component. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
Specifications

Specifications

PIGK
Polyclonal
Unconjugated
PIGK
GPI anchor transamidase, GPI transamidase, GPI8, GPI8 homolog, hGPI8, PIG K, PIGK
Rabbit
Antigen Affinity Chromatography
RUO
10026
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92643
PIGK
PIGK Fusion Protein Ag3497
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers