Learn More
Thermo Scientific™ Pierce™ Power Stainer
Get high quality protein staining with low background in 6 to 11 minutes with the Thermo Scientific™ Pierce™ Power Stainer
Brand:  Thermo Scientific™ 22833EU
Additional Details : Weight : 4.00000kg
 View more versions of this product
Product Code. 15369114
  • 16880.00 NOK / Each
Estimated Shipment date
from Supplier 28-01-2019
Add to basket
Specifications
Specifications
Pierce Power Stainer
Electrophoretic Coomassie staining of proteins in polyacrylamide gels
Benchtop instrument with accessories
Pierce Power Station, Pierce Power Stain Cassette, Western Blot Roller, EU Power Cord with C/13 Connector, Quick Start Guide
2-part unit
Videos