missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphatidyl-L-serine, ∽98%, MP Biomedicals™
Brand: MP Biomedicals 0210096950
Additional Details : Weight : 0.01000kg
Quantity | 50mg |
---|
Chemical Identifiers
51446-62-9 | |
792.089 | |
distearoyl phosphatidylserine, 2s-2-amino-3-r-2,3-bis stearoyloxy propoxy hydroxy phosphoryl oxy propanoic acid, phosphatidylserine 36:0, dioctadecanoylphosphatidylserine, pser 36:0, 1,2-dioctadecanoyl-sn-glycero-3-phosphoserine, phosphatidylserine 18:0/18:0, l-serine, 2,3-bis 1-oxooctadecyl oxy propyl ester, dihydrogen phosphate ester , r, 1,2-distearoyl-sn-glycero-3-phospho-l-serine, l-alpha-phosphatidyl-l-serine | |
CHEBI:84519 | |
CCCCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCC(C(=O)O)N)OC(=O)CCCCCCCCCCCCCCCCC |
C42H82NO10P | |
TZCPCKNHXULUIY-RGULYWFUSA-N | |
9547096 | |
(2S)-2-amino-3-[[(2R)-2,3-di(octadecanoyloxy)propoxy]-hydroxyphosphoryl]oxypropanoic acid |
Specifications
Phosphatidyl-L-serine | |
∽ 98% | |
distearoyl phosphatidylserine, 2s-2-amino-3-r-2,3-bis stearoyloxy propoxy hydroxy phosphoryl oxy propanoic acid, phosphatidylserine 36:0, dioctadecanoylphosphatidylserine, pser 36:0, 1,2-dioctadecanoyl-sn-glycero-3-phosphoserine, phosphatidylserine 18:0/18:0, l-serine, 2,3-bis 1-oxooctadecyl oxy propyl ester, dihydrogen phosphate ester , r, 1,2-distearoyl-sn-glycero-3-phospho-l-serine, l-alpha-phosphatidyl-l-serine | |
CCCCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCC(C(=O)O)N)OC(=O)CCCCCCCCCCCCCCCCC | |
792.089 | |
CHEBI:84519 | |
Liquid | |
50mg |
51446-62-9 | |
C42H82NO10P | |
TZCPCKNHXULUIY-RGULYWFUSA-N | |
(2S)-2-amino-3-[[(2R)-2,3-di(octadecanoyloxy)propoxy]-hydroxyphosphoryl]oxypropanoic acid | |
9547096 | |
788 | |
Yellow |