Learn More
Phenytoin Sodium, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Phenytoin Sodium
Specifications
Specifications
| Chemical Name or Material | Phenytoin sodium |
| CAS | 630-93-3 |
| Analyte or Component Names | Phenytoin sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H11 N2 O2 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Phenytoin Sodium, 5,5-Diphenyl-2,4-imidazolidinedione sodium salt (1:1), 5,5-Diphenyl-2,4-imidazolidinedione monosodium salt, 5,5-Diphenylhydantoin sodium salt, 5,5-Diphenylhydantoin sodium, Aleviatin sodium, Antisacer, Danten, Difenin, Difhydan, Dilantin, Diphantoine, Diphenin, Diphenine, Diphenylan sodium, Diphenylhydantoin sodium, Ditoin, Enkefal, Epanutin, Epelin, Epilan D, Epsolin, Eptoin, Fenitoin sodium, Hydantin, Hydantoinal, M-toin, Minetoin, Phenyloin, Phenytoin sodium, Phenytoin soluble, Prompt, Sodium 5,5-diphenyl-2,4-imidazolidinedione, Sodium 5,5-diphenylhydantoin, Sodium diphenylhydantoin, Sodium diphenylhydantoinate, Sodium phenytoin, Solantyl, Soluble Phenytoin, Tacosal |
| InChI Formula | InChI=1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1 |
| SMILES | [Na+].[O-]C1=NC(=O)C(N1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | sodium;4-oxo-5,5-diphenyl-1H-imidazol-2-olate |
| Show More |